siRNA Details
| Gene Symbol: ZNF263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005741.4 | NP_005732.2 | 2, 3 | 901 | ||
| XM_006720832.1 | XP_006720895.1 | 2, 3 | 1008 | |||
| XM_011522344.1 | XP_011520646.1 | 2, 3 | 1008 | |||
| GenBank mRNA | AK312421.1 | - | 2, 3 | 883 | ||
| BC008805.2 | - | 2, 3 | 899 | 3945548 | ||
| D88827.1 | - | 2, 3 | 861 | |||

ZNF263
Genomic Map