siRNA Details
| Gene Symbol: SLC35G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL136661.1 | - | 2 | 647 | ||
| BC000111.1 | - | 2 | 242 | 3508204 | ||
| BC022557.1 | - | 2 | 287 | 4793991 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC35G2| Gene Symbol: SLC35G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL136661.1 | - | 2 | 647 | ||
| BC000111.1 | - | 2 | 242 | 3508204 | ||
| BC022557.1 | - | 2 | 287 | 4793991 | ||