siRNA Details
| Gene Symbol: RAPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203365.3 | NP_976241.1 | 2 | 299 | ||
| NM_213589.2 | NP_998754.1 | 2 | 299 | |||
| XM_005246777.3 | XP_005246834.1 | 2 | 294 | |||
| XM_005246779.4 | XP_005246836.1 | 2 | 1193 | |||
| XM_006712695.3 | XP_006712758.1 | 3 | 1274 | |||
| XM_011511646.2 | XP_011509948.1 | 3 | 366 | |||
| XM_011511647.1 | XP_011509949.1 | 1 | 74 | |||
| XM_011511648.1 | XP_011509950.1 | 1 | 74 | |||
| XM_011511649.2 | XP_011509951.1 | 2 | 294 | |||
| GenBank mRNA | AB051468.1 | - | 1 | 32 | ||
| AB053311.1 | - | 2 | 293 | |||
| AJ584699.1 | - | 2 | 293 | |||
| AK096270.1 | - | 2 | 243 | |||
| AY494951.1 | - | 2 | 295 | |||
| AY523977.1 | - | 2 | 295 | |||
| AY523978.1 | - | 2 | 295 | |||

RAPH1
Genomic Map