siRNA Details
| Gene Symbol: LY6G6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025261.2 | NP_079537.1 | 2 | 193 | ||
| GenBank mRNA | AJ315533.1 | - | 2 | 138 | ||
| AJ315534.1 | - | 3 | 249 | |||
| AY359119.1 | - | 2 | 193 | |||
| BC036302.1 | - | 2 | 168 | 4776231 | ||

LY6G6C
Genomic Map