siRNA Details
| Gene Symbol: PRR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077497.2 | NP_001070965.1 | 2 | 956 | ||
| NM_025263.3 | NP_079539.2 | 3 | 1019 | |||
| GenBank mRNA | AK074531.1 | - | 2 | 385 | ||
| AK131382.1 | - | 1 | 2347 | |||
| AK298621.1 | - | 3 | 743 | |||
| AK316586.1 | - | 3 | 748 | |||
| BC126455.1 | - | 3 | 599 | 8992171 | ||
| CN258740.1 | - | 3 | 412 | |||
| DR762096.1 | - | 2 | 346 | |||
| U63336.1 | - | 3 | 842 | |||

PRR3
Genomic Map