siRNA Details
| Gene Symbol: REEP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316965.1 | NP_001303894.1 | 6 | 932 | ||
| NM_025232.3 | NP_079508.2 | 7 | 1068 | |||
| GenBank mRNA | AF218014.1 | - | 7 | 846 | ||
| AK025899.1 | - | 7 | 998 | |||
| AK025930.1 | - | 7 | 701 | |||
| AK172846.1 | - | 7 | 1003 | |||
| AY562242.1 | - | 7 | 599 | |||
| BC013048.1 | - | 7 | 1052 | 3863838 | ||
| BC050622.1 | - | 6 | 834 | 6504298 | ||
| CR457353.1 | - | 7 | 599 | |||

REEP4
Genomic Map