siRNA Details
| Gene Symbol: SLC44A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001178044.1 | NP_001171515.1 | 10, 11 | 986 | ||
| NM_001178045.1 | NP_001171516.1 | 11, 12 | 1115 | |||
| NM_025257.2 | NP_079533.2 | 11, 12 | 1112 | |||
| GenBank mRNA | AK027397.1 | - | 11, 12 | 1063 | ||
| AK222998.1 | - | 11, 12 | 1063 | |||
| AK300550.1 | - | 11, 12 | 1115 | |||
| AK301596.1 | - | 10, 11 | 937 | |||
| AK314673.1 | - | 11, 12 | 1067 | |||
| AL833009.1 | - | 7, 8 | 805 | |||
| BC014659.1 | - | 11, 12 | 1051 | 4644272 | ||
| DA924720.1 | - | 4, 5 | 516 | |||
| KU175229.1 | - | 11, 12 | 1046 | |||

SLC44A4
Genomic Map