siRNA Details
Gene Symbol: LY6G6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025261.2 | NP_079537.1 | 3 | 285 | ||
GenBank mRNA | AJ315533.1 | - | 3 | 230 | ||
AJ315534.1 | - | 4 | 341 | |||
AY359119.1 | - | 3 | 285 | |||
BC036302.1 | - | 3 | 260 | 4776231 |
