siRNA Details
| Gene Symbol: NUBPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201573.1 | NP_001188502.1 | 9 | 940 | ||
| NM_001201574.1 | NP_001188503.1 | 6 | 843 | |||
| NM_025152.2 | NP_079428.2 | 11 | 1176 | |||
| NR_120408.1 | 10 | 1082 | ||||
| XM_011537181.2 | XP_011535483.1 | 8 | 1111 | |||
| XM_011537182.2 | XP_011535484.1 | 12 | 1150 | |||
| XM_011537184.2 | XP_011535486.1 | 6 | 1026 | |||
| XM_017021667.1 | XP_016877156.1 | 10 | 1097 | |||
| GenBank mRNA | AK022722.1 | - | 11 | 1089 | ||
| AK093789.1 | - | 6 | 843 | |||
| AK295326.1 | - | 11 | 1137 | |||
| AK301447.1 | - | 9 | 940 | |||
| BC024919.1 | - | 11 | 1102 | 4475196 | ||
| BX248028.1 | - | 10 | 1069 | |||
| BX648419.1 | - | 3 | 3156 | |||

NUBPL
Genomic Map