siRNA Details
| Gene Symbol: TRIM45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145635.1 | NP_001139107.1 | 4 | 1925 | ||
| NM_025188.3 | NP_079464.2 | 4 | 1979 | |||
| XM_011542199.2 | XP_011540501.1 | 5 | 1275 | |||
| GenBank mRNA | AK023243.1 | - | 4 | 1918 | ||
| AK222901.1 | - | 4 | 1923 | |||
| AY669488.1 | - | 4 | 1979 | |||
| BC034943.2 | - | 4 | 1898 | 4826874 | ||

TRIM45
Genomic Map