siRNA Details
| Gene Symbol: EPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272004.1 | NP_001258933.1 | 13, 14 | 2680 | ||
| NM_001272019.2 | NP_001258948.1 | 13, 14 | 2286 | |||
| NM_001282391.1 | NP_001269320.1 | 13, 14 | 2532 | |||
| NM_025209.3 | NP_079485.1 | 14, 15 | 2749 | |||
| XM_006717510.1 | XP_006717573.1 | 14, 15 | 2601 | |||
| GenBank mRNA | AF277374.1 | - | 14, 15 | 2716 | ||
| AF286905.1 | - | 13, 14 | 2350 | |||
| AK024329.1 | - | 10, 11 | 1795 | |||
| AK092304.1 | - | 13, 14 | 2286 | |||
| AK299676.1 | - | 13, 14 | 2473 | |||
| BC036529.1 | - | 13, 14 | 2681 | 4828057 | ||

EPC1
Genomic Map