siRNA Details
| Gene Symbol: THAP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008695.1 | NP_001008695.1 | 4 | 357 | ||
| NM_030573.2 | NP_085050.2 | 3 | 531 | |||
| GenBank mRNA | AK024609.1 | - | 1 | 1095 | ||
| AK025906.1 | - | 1 | 863 | |||
| AK300715.1 | - | 4 | 146 | |||
| AK315457.1 | - | 3 | 516 | |||
| BC004346.1 | - | 3 | 502 | 3633743 | ||
| BE264280.1 | - | 3 | 430 | |||
| BI223500.1 | - | 3 | 474 | |||
| BM790105.1 | - | 4 | 357 | |||
| CR456343.1 | - | 3 | 349 | |||

THAP7
Genomic Map