siRNA Details
| Gene Symbol: FAM49A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030797.3 | NP_110424.1 | 4 | 349 | ||
| XM_005262630.3 | XP_005262687.1 | 3 | 305 | |||
| XM_006712108.3 | XP_006712171.1 | 4 | 304 | |||
| GenBank mRNA | AK315065.1 | - | 3 | 244 | ||
| AL136704.1 | - | 4 | 314 | |||
| BC038971.1 | - | 4 | 300 | 5200807 | ||
| CR533504.1 | - | 2 | 96 | |||
| DA188616.1 | - | 4 | 349 | |||

FAM49A
Genomic Map