siRNA Details
| Gene Symbol: C1orf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030806.3 | NP_110433.1 | 3, 4 | 658 | ||
| GenBank mRNA | AF312864.1 | - | 3, 4 | 580 | ||
| AK056487.1 | - | 3, 4 | 394 | |||
| AK312062.1 | - | 2, 3 | 295 | |||
| AY258286.1 | - | 3, 4 | 571 | |||
| BC028567.2 | - | 3, 4 | 616 | 3635521 | ||

C1orf21
Genomic Map