siRNA Details
| Gene Symbol: CREBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310.3 | NP_001301.1 | 4 | 874 | ||
| GenBank mRNA | AF039081.1 | - | 4 | 810 | ||
| AK307202.1 | - | 5 | 757 | |||
| BC104873.1 | - | 4 | 718 | 8143876 | ||
| BC106052.1 | - | 4 | 815 | 6150812 | ||
| BC112157.1 | - | 4 | 718 | 8327625 | ||

CREBL2
Genomic Map