siRNA Details
| Gene Symbol: LOC101929066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | N58047.1 | - | 1 | 73 | ||
| Gene Symbol: ZNF696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030895.2 | NP_112157.2 | 3 | 2599 | ||
| GenBank mRNA | AK024191.1 | - | 3 | 2563 | ||

LOC101929066
Genomic Map