siRNA Details
Gene Symbol: LIMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030576.3 | NP_085053.1 | 5 | 757 | ||
XM_005257703.1 | XP_005257760.1 | 5 | 688 | |||
XM_005257705.3 | XP_005257762.1 | 5 | 868 | |||
XM_006722124.1 | XP_006722187.1 | 5 | 664 | |||
GenBank mRNA | AK092301.1 | - | 5 | 723 | ||
AK307353.1 | - | 3 | 733 | |||
AK310718.1 | - | 2 | 819 | |||
AY037154.1 | - | 5 | 812 | |||
BC004400.1 | - | 5 | 738 | 3636469 | ||
BC051812.1 | - | 5 | 993 | 5732102 |
