siRNA Details
| Gene Symbol: CLPTM1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030782.4 | NP_110409.2 | 5, 6 | 906 | ||
| XM_011514144.2 | XP_011512446.1 | 5, 6 | 870 | |||
| GenBank mRNA | AB045223.1 | - | 5, 6 | 663 | ||
| AK027306.1 | - | 5, 6 | 720 | |||
| BC016399.1 | - | 3, 4 | 292 | |||
| BC025305.1 | - | 5, 6 | 724 | 3051368 | ||

CLPTM1L
Genomic Map