siRNA Details
| Gene Symbol: SLC25A21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171170.1 | NP_001164641.1 | 8, 9 | 1102 | ||
| NM_030631.3 | NP_085134.1 | 8, 9 | 1102 | |||
| XM_011537287.2 | XP_011535589.1 | 9, 10 | 884 | |||
| XM_011537288.2 | XP_011535590.1 | 8, 9 | 976 | |||
| XM_011537289.2 | XP_011535591.1 | 7, 8 | 885 | |||
| GenBank mRNA | AJ278148.1 | - | 8, 9 | 1102 | ||
| AK289575.1 | - | 8, 9 | 865 | |||
| BC101521.1 | - | 8, 9 | 838 | 8069027 | ||
| BC113365.1 | - | 8, 9 | 838 | 8322417 | ||

SLC25A21
Genomic Map