siRNA Details
| Gene Symbol: ACAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030649.2 | NP_085152.2 | 6 | 439 | ||
| XM_005244715.2 | XP_005244772.1 | 6 | 475 | |||
| XM_011540606.2 | XP_011538908.1 | 6 | 488 | |||
| XM_011540607.1 | XP_011538909.1 | 6 | 2912 | |||
| XM_011540608.2 | XP_011538910.1 | 6 | 489 | |||
| XM_011540609.2 | XP_011538911.1 | 6 | 489 | |||
| XM_017000233.1 | XP_016855722.1 | 6 | 628 | |||
| GenBank mRNA | AB051503.1 | - | 5 | 2104 | ||
| AF411981.1 | - | 4 | 239 | |||
| AK093954.1 | - | 6 | 486 | |||
| BC051194.1 | - | 6 | 439 | 4819254 | ||

ACAP3
Genomic Map