siRNA Details
| Gene Symbol: CFHR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030787.3 | NP_110414.1 | 5 | 796 | ||
| XM_011510020.2 | XP_011508322.1 | 5 | 703 | |||
| GenBank mRNA | AF295327.1 | - | 5 | 761 | ||
| AK314026.1 | - | 5 | 797 | |||
| BC026282.1 | - | 5 | 777 | |||
| BC111773.1 | - | 5 | 761 | 40034462 | ||

CFHR5
Genomic Map