siRNA Details
| Gene Symbol: ZNF696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030895.2 | NP_112157.2 | 3 | 2646 | ||
| GenBank mRNA | AK024191.1 | - | 3 | 2610 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF696| Gene Symbol: ZNF696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030895.2 | NP_112157.2 | 3 | 2646 | ||
| GenBank mRNA | AK024191.1 | - | 3 | 2610 | ||