siRNA Details
| Gene Symbol: PABPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030979.2 | NP_112241.2 | 1 | 1194 | ||
| GenBank mRNA | AF132026.1 | - | 1 | 1156 | ||
| AK313336.1 | - | 1 | 1176 | |||
| AL136900.1 | - | 1 | 1176 | |||
| BC027617.2 | - | 1 | 1160 | 4829121 | ||
| BC045608.1 | - | 1 | 1170 | 5273503 | ||

PABPC3
Genomic Map