siRNA Details
Gene Symbol: TEX13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031273.2 | NP_112563.1 | 2 | 384 | ||
GenBank mRNA | AF285598.1 | - | 2 | 384 | ||
BC150140.1 | - | 1 | 523 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TEX13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031273.2 | NP_112563.1 | 2 | 384 | ||
GenBank mRNA | AF285598.1 | - | 2 | 384 | ||
BC150140.1 | - | 1 | 523 |