siRNA Details
| Gene Symbol: TDRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198795.1 | NP_942090.1 | 10, 11 | 1458 | ||
| XM_005269978.2 | XP_005270035.1 | 10, 11 | 1439 | |||
| XM_005269979.2 | XP_005270036.1 | 10, 11 | 1472 | |||
| XM_011539959.2 | XP_011538261.1 | 10, 11 | 1811 | |||
| XM_011539960.2 | XP_011538262.1 | 10, 11 | 1401 | |||
| XM_011539961.2 | XP_011538263.1 | 10, 11 | 1383 | |||
| XM_011539962.1 | XP_011538264.1 | 10, 11 | 1765 | |||
| XM_011539963.1 | XP_011538265.1 | 10, 11 | 1472 | |||
| XM_011539964.1 | XP_011538266.1 | 10, 11 | 1472 | |||
| XM_011539966.1 | XP_011538268.1 | 10, 11 | 1472 | |||
| XM_017016414.1 | XP_016871903.1 | 9, 10 | 1328 | |||
| XM_017016415.1 | XP_016871904.1 | 9, 10 | 1328 | |||
| GenBank mRNA | AF285606.1 | - | 3, 4 | 427 | ||
| AK122916.1 | - | 10, 11 | 1468 | |||
| AK304331.1 | - | 2, 3 | 426 | |||
| AL833340.1 | - | 10, 11 | 1460 | |||
| BC035010.2 | - | 10, 11 | 1764 | |||
| BC063133.1 | - | 10, 11 | 1366 | 6710928 | ||
| BX648664.1 | - | 10, 11 | 1403 | |||

TDRD1
Genomic Map