siRNA Details
| Gene Symbol: RAB33B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031296.2 | NP_112586.1 | 2 | 668 | ||
| XM_011532299.1 | XP_011530601.1 | 3 | 764 | |||
| GenBank mRNA | AF350420.1 | - | 2 | 390 | ||
| AK313685.1 | - | 2 | 328 | |||
| AL136904.1 | - | 2 | 668 | |||
| BC036064.1 | - | 2 | 300 | 5288066 | ||
| BC111977.1 | - | 2 | 320 | 8327445 | ||

RAB33B
Genomic Map