siRNA Details
| Gene Symbol: NECTIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030916.2 | NP_112178.2 | 4 | 1100 | ||
| XM_005245508.3 | XP_005245565.1 | 4 | 1071 | |||
| XM_011510021.2 | XP_011508323.1 | 4 | 1071 | |||
| XM_011510022.2 | XP_011508324.1 | 4 | 1071 | |||
| XM_011510023.1 | XP_011508325.1 | 4 | 1071 | |||
| GenBank mRNA | AB105191.1 | - | 4 | 1057 | ||
| AB755430.1 | - | 4 | 801 | |||
| AF160477.1 | - | 4 | 936 | |||
| AF426163.1 | - | 4 | 801 | |||
| AK027753.1 | - | 4 | 1057 | |||
| AK298981.1 | - | 1 | 78 | |||
| BC010423.1 | - | 4 | 1048 | 3610849 | ||

NECTIN4
Genomic Map