siRNA Details
| Gene Symbol: UNC93B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030930.3 | NP_112192.2 | 3, 4 | 482 | ||
| XM_011545291.2 | XP_011543593.1 | 2, 3 | 304 | |||
| GenBank mRNA | AJ271326.1 | - | 3, 4 | 441 | ||
| AJ422142.1 | - | 3, 4 | 450 | |||
| AY007125.1 | - | 2, 3 | 269 | |||
| BC025669.1 | - | 3, 4 | 428 | |||
| BC033623.1 | - | 3, 4 | 480 | |||
| BC092472.1 | - | 3, 4 | 450 | 30348121 | ||
| BC101568.1 | - | 3, 4 | 426 | 8069074 | ||
| BC105104.1 | - | 3, 4 | 426 | 8144107 | ||

UNC93B1
Genomic Map