siRNA Details
| Gene Symbol: SPRY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127496.1 | NP_001120968.1 | 2 | 2594 | ||
| NM_001293289.1 | NP_001280218.1 | 3 | 2972 | |||
| NM_001293290.1 | NP_001280219.1 | 2 | 2910 | |||
| NM_030964.3 | NP_112226.2 | 3 | 2645 | |||
| XM_011537685.2 | XP_011535987.1 | 3 | 3372 | |||
| XM_017009910.1 | XP_016865399.1 | 2 | 3638 | |||
| GenBank mRNA | AF227516.1 | - | 3 | 2586 | ||
| AK226147.1 | - | 1 | 3017 | |||

SPRY4
Genomic Map