siRNA Details
| Gene Symbol: KRTAP1-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030966.1 | NP_112228.1 | 1 | 933 | ||
| GenBank mRNA | AJ406927.1 | - | 1 | 933 | ||
| BC069161.1 | - | 1 | 932 | 7216931 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP1-3| Gene Symbol: KRTAP1-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030966.1 | NP_112228.1 | 1 | 933 | ||
| GenBank mRNA | AJ406927.1 | - | 1 | 933 | ||
| BC069161.1 | - | 1 | 932 | 7216931 | ||