siRNA Details
| Gene Symbol: PSG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | M20882.1 | - | 4 | 1021 | ||
| M37398.1 | - | 1 | 211 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PSG2| Gene Symbol: PSG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | M20882.1 | - | 4 | 1021 | ||
| M37398.1 | - | 1 | 211 | |||