siRNA Details
| Gene Symbol: CCDC70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031290.2 | NP_112580.2 | 2 | 493 | ||
| GenBank mRNA | AK098719.1 | - | 2 | 493 | ||
| AK225745.1 | - | 2 | 466 | |||
| AL136887.1 | - | 2 | 464 | |||
| BC069691.1 | - | 2 | 336 | 7262643 | ||
| BC069748.1 | - | 2 | 336 | 7262655 | ||
| BC069770.1 | - | 2 | 336 | 7262572 | ||
| BC069806.1 | - | 2 | 336 | 7262584 | ||
| BP371671.1 | - | 2 | 493 | |||

CCDC70
Genomic Map