siRNA Details
| Gene Symbol: DLX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005222.3 | NP_005213.3 | 2 | 498 | ||
| GenBank mRNA | AK094086.1 | - | 2 | 309 | ||
| BC069363.1 | - | 2 | 144 | 7262091 | ||
| BC103688.1 | - | 2 | 177 | 40000523 | ||
| BC103689.1 | - | 2 | 177 | 40000521 | ||
| BC103690.1 | - | 2 | 177 | 40000520 | ||
| BC109381.1 | - | 2 | 177 | 40000518 | ||

DLX6
Genomic Map