siRNA Details
| Gene Symbol: SHCBP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030933.2 | NP_112195.2 | 7 | 1481 | ||
| GenBank mRNA | AF288397.1 | - | 8 | 1139 | ||
| AF288398.1 | - | 7 | 1480 | |||
| BC026084.1 | - | 6 | 1661 | |||
| BC050305.1 | - | 7 | 1433 | 4837018 | ||
| BC132764.1 | - | 7 | 1457 | 40146786 | ||
| BC144326.1 | - | 7 | 1453 | |||

SHCBP1L
Genomic Map