siRNA Details
| Gene Symbol: LINC00597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026813.1 | 1 | 782 | |||
| GenBank mRNA | AF216224.1 | - | 1 | 788 | ||
| AL109679.1 | - | 1 | 427 | |||
| BC069765.1 | - | 1 | 718 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00597| Gene Symbol: LINC00597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026813.1 | 1 | 782 | |||
| GenBank mRNA | AF216224.1 | - | 1 | 788 | ||
| AL109679.1 | - | 1 | 427 | |||
| BC069765.1 | - | 1 | 718 | |||