siRNA Details
| Gene Symbol: SPACA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030960.2 | NP_112222.1 | 1, 2 | 329 | ||
| GenBank mRNA | AF203447.1 | - | 1, 2 | 284 | ||
| BC029488.1 | - | 1, 2 | 329 | 5270534 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPACA1| Gene Symbol: SPACA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030960.2 | NP_112222.1 | 1, 2 | 329 | ||
| GenBank mRNA | AF203447.1 | - | 1, 2 | 284 | ||
| BC029488.1 | - | 1, 2 | 329 | 5270534 | ||