siRNA Details
| Gene Symbol: ST6GALNAC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320273.1 | NP_001307202.1 | 3 | 606 | ||
| NM_030965.2 | NP_112227.1 | 4 | 1016 | |||
| GenBank mRNA | AJ507292.1 | - | 4 | 760 | ||
| AK056241.1 | - | 4 | 983 | |||
| AK095758.1 | - | 5 | 902 | |||
| AK295019.1 | - | 5 | 859 | |||
| AK300902.1 | - | 3 | 531 | |||
| BC001201.1 | - | 4 | 923 | 3356535 | ||

ST6GALNAC5
Genomic Map