siRNA Details
| Gene Symbol: MRPS15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031280.3 | NP_112570.2 | 7 | 660 | ||
| GenBank mRNA | AB049946.1 | - | 7 | 535 | ||
| AB209905.1 | - | 5 | 4813 | |||
| AF265439.1 | - | 7 | 579 | |||
| AK021626.1 | - | 3 | 1320 | |||
| AK125230.1 | - | 2 | 1834 | |||
| AK315441.1 | - | 7 | 599 | |||
| BC031336.1 | - | 7 | 575 | 3543896 | ||
| BM048518.1 | - | 7 | 659 | |||
| BU927793.1 | - | 7 | 510 | |||
| CF123360.1 | - | 1 | 65 | |||

MRPS15
Genomic Map