siRNA Details
| Gene Symbol: STAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005419.3 | NP_005410.1 | 21 | 2170 | ||
| NM_198332.1 | NP_938146.1 | 21 | 2158 | |||
| XM_011538697.2 | XP_011536999.1 | 21 | 2102 | |||
| XM_011538698.2 | XP_011537000.1 | 21 | 2063 | |||
| XM_011538700.2 | XP_011537002.1 | 15 | 1500 | |||
| XM_017019904.1 | XP_016875393.1 | 20 | 2537 | |||
| GenBank mRNA | AK296939.1 | - | 21 | 2031 | ||
| BC051284.1 | - | 20 | 1990 | 6460486 | ||
| BX640607.1 | - | 8 | 897 | |||
| M97934.1 | - | 21 | 2004 | |||

STAT2
Genomic Map