siRNA Details
| Gene Symbol: EOMES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278182.1 | NP_001265111.1 | 6 | 2518 | ||
| NM_001278183.1 | NP_001265112.1 | 6 | 1502 | |||
| NM_005442.3 | NP_005433.2 | 6 | 2461 | |||
| XM_005265510.3 | XP_005265567.1 | 7 | 2397 | |||
| GenBank mRNA | AB031038.1 | - | 6 | 2040 | ||
| AK297389.1 | - | 7 | 1518 | |||
| AK297461.1 | - | 4 | 2171 | |||
| AK299499.1 | - | 7 | 1900 | |||
| AK299695.1 | - | 7 | 1584 | |||
| AK316166.1 | - | 6 | 1502 | |||
| BC025363.1 | - | 5 | 1154 | |||
| BC037568.1 | - | 6 | 2121 | |||

EOMES
Genomic Map