siRNA Details
| Gene Symbol: CCDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031455.3 | NP_113643.1 | 1 | 490 | ||
| GenBank mRNA | AL136562.1 | - | 1 | 406 | ||
| BC051334.1 | - | 1 | 490 | 6302570 | ||
| DN996248.1 | - | 1 | 485 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC3| Gene Symbol: CCDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031455.3 | NP_113643.1 | 1 | 490 | ||
| GenBank mRNA | AL136562.1 | - | 1 | 406 | ||
| BC051334.1 | - | 1 | 490 | 6302570 | ||
| DN996248.1 | - | 1 | 485 | |||