siRNA Details
| Gene Symbol: INHBE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031479.3 | NP_113667.1 | 1 | 502 | ||
| GenBank mRNA | AF412024.1 | - | 1 | 486 | ||
| AK075285.1 | - | 1 | 497 | |||
| BC005161.2 | - | 1 | 494 | 3529092 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

INHBE| Gene Symbol: INHBE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031479.3 | NP_113667.1 | 1 | 502 | ||
| GenBank mRNA | AF412024.1 | - | 1 | 486 | ||
| AK075285.1 | - | 1 | 497 | |||
| BC005161.2 | - | 1 | 494 | 3529092 | ||