siRNA Details
| Gene Symbol: CHST9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031422.5 | NP_113610.2 | 5 | 529 | ||
| XM_006722555.3 | XP_006722618.1 | 5 | 486 | |||
| XM_017026033.1 | XP_016881522.1 | 4 | 428 | |||
| XM_017026034.1 | XP_016881523.1 | 6 | 652 | |||
| GenBank mRNA | AF239821.1 | - | 5 | 406 | ||
| AF332472.1 | - | 4 | 320 | |||
| AK093349.1 | - | 4 | 428 | |||
| AY358488.1 | - | 5 | 508 | |||
| BC025764.1 | - | 5 | 473 | 5205829 | ||
| KU178792.1 | - | 4 | 215 | |||

CHST9
Genomic Map