siRNA Details
| Gene Symbol: DNAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201366.1 | NP_001188295.1 | 7 | 547 | ||
| NM_031427.3 | NP_113615.2 | 6 | 501 | |||
| XM_017021679.1 | XP_016877168.1 | 7 | 535 | |||
| GenBank mRNA | AF542071.1 | - | 6 | 377 | ||
| AI242197.1 | - | 4 | 352 | |||
| AK026653.1 | - | 3 | 159 | |||
| AK315392.1 | - | 6 | 396 | |||
| AL833654.1 | - | 5 | 257 | |||
| BC005343.1 | - | 6 | 377 | 3930213 | ||
| BJ996719.1 | - | 2 | 97 | |||

DNAL1
Genomic Map