siRNA Details
| Gene Symbol: CRISPLD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031461.5 | NP_113649.1 | 2 | 682 | ||
| GenBank mRNA | AF142573.1 | - | 2 | 278 | ||
| AF329197.1 | - | 1 | 257 | |||
| AK314048.1 | - | 2 | 438 | |||
| AL834301.1 | - | 2 | 414 | |||
| AY358564.1 | - | 2 | 640 | |||
| BC020514.1 | - | 2 | 547 | 3856615 | ||

CRISPLD1
Genomic Map