siRNA Details
| Gene Symbol: CDCA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297602.1 | NP_001284531.1 | 6 | 1726 | ||
| NM_001297604.1 | NP_001284533.1 | 5 | 1694 | |||
| NM_031299.5 | NP_112589.1 | 6 | 1801 | |||
| NR_123727.1 | 6 | 1785 | ||||
| GenBank mRNA | AK291421.1 | - | 6 | 1004 | ||
| BC002551.2 | - | 6 | 996 | 3138951 | ||

CDCA3
Genomic Map