siRNA Details
| Gene Symbol: NFKBIZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005474.2 | NP_001005474.1 | 13 | 2534 | ||
| NM_031419.3 | NP_113607.1 | 12 | 2667 | |||
| GenBank mRNA | AB037925.1 | - | 12 | 2600 | ||
| AF548362.1 | - | 13 | 2531 | |||
| BC014587.2 | - | 2583 | ||||
| BC060800.1 | - | 12 | 2667 | 30344780 | ||

NFKBIZ
Genomic Map