siRNA Details
| Gene Symbol: DNAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201366.1 | NP_001188295.1 | 9 | 1200 | ||
| NM_031427.3 | NP_113615.2 | 8 | 1154 | |||
| XM_017021679.1 | XP_016877168.1 | 9 | 1188 | |||
| GenBank mRNA | AL833654.1 | - | 7 | 909 | ||
| BC005343.1 | - | 8 | 1031 | 3930213 | ||

DNAL1
Genomic Map