siRNA Details
| Gene Symbol: ZBTB16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018011.1 | NP_001018011.1 | 7 | 2203 | ||
| NM_006006.4 | NP_005997.2 | 7 | 2358 | |||
| XM_017018257.1 | XP_016873746.1 | 7 | 2291 | |||
| XM_017018258.1 | XP_016873747.1 | 7 | 2446 | |||
| XM_017018259.1 | XP_016873748.1 | 7 | 2798 | |||
| GenBank mRNA | BC026902.1 | - | 7 | 2262 | 4944546 | |
| BC029812.1 | - | 7 | 2340 | 5175961 | ||
| BX648973.1 | - | 7 | 2475 | |||
| Z19002.1 | - | 6 | 2169 | |||

ZBTB16
Genomic Map