siRNA Details
| Gene Symbol: TEX101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130011.1 | NP_001123483.1 | 5 | 702 | ||
| NM_031451.4 | NP_113639.4 | 8 | 1122 | |||
| XM_005259303.3 | XP_005259360.2 | 5 | 692 | |||
| GenBank mRNA | AF241268.1 | - | 8 | 1121 | ||
| AF353396.1 | - | 5 | 792 | |||
| AY014285.1 | - | 5 | 674 | |||
| AY359077.1 | - | 5 | 670 | |||
| BC001861.2 | - | 6 | 869 | 3538494 | ||

TEX101
Genomic Map